Oxalic acid bis(methyl 4-{[(tert-butoxy)carbonyl]amino}piperidine-4-carboxylate) ≥98.5%

N20120686-01-1 N20120686-01-2 N20120686-01-3
103536-478EA 92.31 USD
103536-478 103536-480 103536-482
Oxalic acid bis(methyl 4-{[(tert-butoxy)carbonyl]amino}piperidine-4-carboxylate) ≥98.5%
Oxalic acid bis(methyl 4-{[(tert-butoxy)carbonyl]amino}piperidine-4-carboxylate)
Formula: C₂₆H₄₆N₄O₁₂
MW: 606.663 g/mol
CAS Number: 1788043-91-3

Order Now

Specification Test Results

Purety 98.5%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR