(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane ≥99.3%

Synonyms: tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate

N20120579-1 N20120579-2 N20120579-3
103536-466EA 84.62 USD
103536-466 103536-468 103536-470
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane ≥99.3%
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane
Formula: C₁₀H₁₈N₂O₂
MW: 198.26 g/mol
MDL Number: MFCD01569250
CAS Number: 113451-59-5

Order Now

Specification Test Results

Purety 99.3%
Appearance White Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR