2,6-Dichloro-3-cyanopyrazine ≥99.9%

Synonyms: 3,5-dichloropyrazine-2-carbonitrile

N20120534-1 N20120534-2 N20120534-3
103536-460EA 61.54 USD
103536-460 103536-462 103536-464
2,6-Dichloro-3-cyanopyrazine ≥99.9%
2,6-Dichloro-3-cyanopyrazine
Formula: C₅HCl₂N₃
MW: 173.9875 g/mol
MDL Number: MFCD13193460
CAS Number: 313339-92-3

Order Now

Specification Test Results

Purety 99.9%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR