(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid ≥97.0%

Synonyms: (2S)-1-[(tert-butoxy)carbonyl]-4,4-difluoropyrrolidine-2-carboxylic acid, (S)-1-(tert-Butoxycarbonyl)-4,4-difluoropyrrolidine-2-carboxylic acid, N-Boc-4,4-difluoro-L-proline, (S)-1-Boc-4,4-difluoro-2-pyrrolidinecarboxylic Acid, N-(tert-Butoxycarbonyl)-4,4-difluoro-L-proline

N20120406-1 N20120406-2 N20120406-3
103536-448EA 69.23 USD
103536-448 103536-450 103536-452
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid ≥97.0%
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid
Formula: C₁₀H₁₅F₂NO₄
MW: 251.23 g/mol
MDL Number: MFCD03094917
CAS Number: 203866-15-3

Order Now

Specification Test Results

Purety 97.0%
Appearance off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR