7-Fluoro-6-nitroquinazolin-4(3H)-one ≥97.0%

Synonyms: 7-Fluoro-6-nitroquinazolin-4(1H)-one, 7-fluoro-6-nitro-3,4-dihydroquinazolin-4-one, 7-Fluoro-6-nitroquinazolin-4(3H)-one, 7-Fluoro-4-hydroxy-6-nitroquinazoline

N20120356-1 N20120356-2 N20120356-3
103536-436EA 30.77 USD
103536-436 103536-438 103536-440
7-Fluoro-6-nitroquinazolin-4(3H)-one ≥97.0%
7-Fluoro-6-nitroquinazolin-4(3H)-one
Formula: C₈H₄FN₃O₃
MW: 209.1300 g/mol
MDL Number: MFCD08690890
CAS Number: 162012-69-3

Order Now

Specification Test Results

Purety 97.0%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR