4-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole ≥99.2%

Synonyms: 4-bromo-1-(oxan-2-yl)-1H-indazole

N20120246-1 N20120246-2 N20120246-3
103536-430EA 230.77 USD
103536-430 103536-432 103536-434
4-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole ≥99.2%
4-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole
Formula: C₁₂H₁₃BrN₂O
MW: 281.16 g/mol
MDL Number: MFCD22380249
CAS Number: 1022158-35-5

Order Now

Specification Test Results

Purety 99.2%
Appearance White Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR