Ethyl-2,4-dichloropyrimidine-5-carboxylate ≥97.0%

Synonyms: Ethyl 2,4-Dichloro-5-pyrimidinecarboxylate

N20120214-1 N20120214-2 N20120214-3
103535-568EA 30.77 USD
103535-568 103536-426 103536-428
Ethyl-2,4-dichloropyrimidine-5-carboxylate ≥97.0%
Ethyl-2,4-dichloropyrimidine-5-carboxylate
Formula: C₇H₆Cl₂N₂O₂
MW: 221.04 g/mol
MDL Number: MFCD09910281
CAS Number: 51940-64-8

Order Now

Specification Test Results

Purety 97.0%
Appearance Off-white Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR