2-Chloro-5-methylpyrimidine ≥99.4%

N20120211-1 N20120211-2 N20120211-3
103535-562EA 12.31 USD
103535-562 103535-564 103535-566
2-Chloro-5-methylpyrimidine ≥99.4%
2-Chloro-5-methylpyrimidine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
MDL Number: MFCD09260903
CAS Number: 22536-61-4

Order Now

Specification Test Results

Purety 99.4%
Appearance light brown powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR