2-Chloro-4-methylpyrimidine ≥98.6%

N20120175-1 N20120175-2 N20120175-3
103535-556EA 30.77 USD
103535-556 103535-558 103535-560
2-Chloro-4-methylpyrimidine ≥98.6%
2-Chloro-4-methylpyrimidine
Formula: C₅H₅ClN₂
MDL Number: MFCD00054434
CAS Number: 13036-57-2

Order Now

Specification Test Results

Purety 98.6%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR