4-Amino-5-iodopyrrolo[2,3-d]pyrimidine ≥97.1%

N20120170-1 N20120170-2 N20120170-3
103535-550EA 69.23 USD
103535-550 103535-552 103535-554
4-Amino-5-iodopyrrolo[2,3-d]pyrimidine ≥97.1%
4-Amino-5-iodopyrrolo[2,3-d]pyrimidine
Formula: C₆H₅IN₄
MDL Number: MFCD13619882
CAS Number: 163622-50-2

Order Now

Specification Test Results

Purety 97.1%
Appearance Gray solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR