3,3-Dimethoxycyclobutane-1-carboxylate methyl ester ≥99.2%

Synonyms: Methyl 3,3-dimethoxycyclobutanecarboxylate, methyl 3,3-dimethoxycyclobutane-1-carboxylate

N20120162-1 N20120162-2 N20120162-3
103535-544EA 30.77 USD
103535-544 103535-546 103535-548
3,3-Dimethoxycyclobutane-1-carboxylate methyl ester ≥99.2%
3,3-Dimethoxycyclobutane-1-carboxylate methyl ester
Formula: C₈H₁₄O₄
MW: 174.2 g/mol
MDL Number: MFCD11111944
CAS Number: 98231-07-3

Order Now

Specification Test Results

Purety 99.2%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR