Diisopropyl-3,3-difluorocyclobutane-1,1-dicarboxylate ≥97.1%

Synonyms: 1,1-bis(propan-2-yl) 3,3-difluorocyclobutane-1,1-dicarboxylate

N20120145-1 N20120145-2 N20120145-3
103535-538EA 184.62 USD
103535-538 103535-540 103535-542
Diisopropyl-3,3-difluorocyclobutane-1,1-dicarboxylate ≥97.1%
Diisopropyl-3,3-difluorocyclobutane-1,1-dicarboxylate
Formula: C₁₂H₁₈F₂O₄
MW: 264.27 g/mol
MDL Number: MFCD23105827
CAS Number: 1225532-89-7

Order Now

Specification Test Results

Purety 97.1%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR