2,9-Diazaspiro[5.5]undecane-9-carboxylic acid tert-butyl ester ≥98.9%

Synonyms: tert-butyl 2,9-diazaspiro[5.5]undecane-9-carboxylate, 9-Boc-2,9-diazaspiro[5.5]undecane

N2011967-1 N2011967-2 N2011967-3
103535-532EA 100 USD
103535-532 103535-534 103535-536
2,9-Diazaspiro[5.5]undecane-9-carboxylic acid tert-butyl ester ≥98.9%
2,9-Diazaspiro[5.5]undecane-9-carboxylic acid tert-butyl ester
Formula: C₁₄H₂₆N₂O₂
MW: 254.38 g/mol
MDL Number: MFCD10700117
CAS Number: 1023595-19-8

Order Now

Specification Test Results

Purety 98.9%
Appearance Light-Yellow oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR