trans-4-Oxo-1,2-cyclohexanedicarboxylic acid dimethyl ester ≥97%

Synonyms: trans-Dimethyl 4-oxocyclohexane-1,2-dicarboxylate

N2011798-1 N2011798-2 N2011798-3
103535-520EA 30.77 USD
103535-520 103535-522 103535-524
trans-4-Oxo-1,2-cyclohexanedicarboxylic acid dimethyl ester ≥97%
trans-4-Oxo-1,2-cyclohexanedicarboxylic acid dimethyl ester
Formula: C₁₀H₁₄O₅
MW: 214.215 g/mol
MDL Number: MFCD20926192
CAS Number: 13991-44-1

Order Now

Specification Test Results

Purety 97%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR