3-Benzyloxycyclobutanecarboxylic acid ≥97.0%

Synonyms: 3-(benzyloxy)cyclobutane-1-carboxylic acid, 3-(Benzyloxy)cyclobutanecarboxylic acid

N2011746-1 N2011746-2 N2011746-3
103535-514EA 46.15 USD
103535-514 103535-516 103535-518
3-Benzyloxycyclobutanecarboxylic acid ≥97.0%
3-Benzyloxycyclobutanecarboxylic acid
Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
MDL Number: MFCD00512751
CAS Number: 4958-02-5

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR