tert-Butyl 2-oxo-6-azaspiro[3.4]octane-6-carboxylate ≥99.1%

Synonyms: 6-Azaspiro[3.4]octan-2-one, N-BOC protected

N2011625-1 N2011625-2 N2011625-3
103535-508EA 184.62 USD
103535-508 103535-510 103535-512
tert-Butyl 2-oxo-6-azaspiro[3.4]octane-6-carboxylate ≥99.1%
tert-Butyl 2-oxo-6-azaspiro[3.4]octane-6-carboxylate
Formula: C₁₂H₁₉NO₃
MW: 225.284 g/mol
MDL Number: MFCD17016202
CAS Number: 203661-71-6

Order Now

Specification Test Results

Purety 99.1%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR