7-(tert-Butoxycarbonyl)-7-azaspiro[3.5]nonane-2-carboxylic acid ≥97.0%

Synonyms: 7-[(tert-butoxy)carbonyl]-7-azaspiro[3.5]nonane-2-carboxylic acid

N2011606-1 N2011606-2 N2011606-3
103535-502EA 230.77 USD
103535-502 103535-504 103535-506
7-(tert-Butoxycarbonyl)-7-azaspiro[3.5]nonane-2-carboxylic acid ≥97.0%
7-(tert-Butoxycarbonyl)-7-azaspiro[3.5]nonane-2-carboxylic acid
Formula: C₁₄H₂₃NO₄
MW: 269.34 g/mol
MDL Number: MFCD12198556
CAS Number: 873924-12-0

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR