(R)-(-)-Prolinol ≥97.1%

Synonyms: D-Prolinol, (2R)-pyrrolidin-2-ylmethanol, D(-)-Prolinol

N2011412-2 N2011412-3 N2011412-1
103535-492EA 38.46 USD
103535-492 103535-494 103536-424
(R)-(-)-Prolinol ≥97.1%
(R)-(-)-Prolinol
Formula: C₅H₁₁NO
MW: 101.1 g/mol
MDL Number: MFCD00064321
CAS Number: 68832-13-3

Order Now

Specification Test Results

Purety 97.1%
Appearance Light-Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR