4,4-Difluorocyclohexanecarboxylic acid ≥99.1%

Synonyms: 4,4-difluorocyclohexane-1-carboxylic acid

N2011394-1 N2011394-2 N2011394-3
103536-418EA 30.77 USD
103536-418 103536-420 103536-422
4,4-Difluorocyclohexanecarboxylic acid ≥99.1%
4,4-Difluorocyclohexanecarboxylic acid
Formula: C₇H₁₀F₂O₂
MW: 164.15 g/mol
MDL Number: MFCD03788493
CAS Number: 122665-97-8

Order Now

Specification Test Results

Purety 99.1%
Appearance White State

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR