2-Bromo-5-nitro-4-picoline ≥97.0%

Synonyms: 2-Bromo-4-methyl-5-nitropyridine

N2011274-1 N2011274-2 N2011274-3
103536-406EA 38.46 USD
103536-406 103536-408 103536-410
2-Bromo-5-nitro-4-picoline ≥97.0%
2-Bromo-5-nitro-4-picoline
Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
MDL Number: MFCD03095066
CAS Number: 23056-47-5

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR