5-Bromopyrazolo[1,5-a]pyridine ≥97.2%

N2011257-1 N2011257-2 N2011257-3
103536-400EA 315.38 USD
103536-400 103536-402 103536-404
5-Bromopyrazolo[1,5-a]pyridine ≥97.2%
5-Bromopyrazolo[1,5-a]pyridine
Formula: C₇H₅BrN₂
MW: 197.032 g/mol
MDL Number: MFCD13189494
CAS Number: 1060812-84-1

Order Now

Specification Test Results

Purety 97.2%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR