tert-Butyl exo-3-amino-9-azabicyclo[3.3.1]nonane-9-carboxylate ≥97%

N2011250-1 N2011250-2 N2011250-3
103536-394EA 523.08 USD
103536-394 103536-396 103536-398
tert-Butyl exo-3-amino-9-azabicyclo[3.3.1]nonane-9-carboxylate ≥97%
tert-Butyl exo-3-amino-9-azabicyclo[3.3.1]nonane-9-carboxylate
Formula: C₁₃H₂₄N₂O₂
CAS Number: 1363380-67-9

Order Now

Specification Test Results

Purety 97%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR