Diethyl fluoromalonate ≥97.0%

Synonyms: 1,3-diethyl 2-fluoropropanedioate

N2011163-1 N2011163-2 N2011163-3
103536-382EA 18.46 USD
103536-382 103536-384 103536-386
Diethyl fluoromalonate ≥97.0%
Diethyl fluoromalonate
Formula: C₇H₁₁FO₄
MW: 178.16 g/mol
MDL Number: MFCD00009139
CAS Number: 685-88-1

Order Now

Specification Test Results

Purety 97.0%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR