Ethyl 6-bromopyrazolo[1,5-a]pyridine-3-carboxylate ≥97%

N2011096-1 N2011096-2 N2011096-3
103535-484EA 184.62 USD
103535-484 103535-486 103535-488
Ethyl 6-bromopyrazolo[1,5-a]pyridine-3-carboxylate ≥97%
Ethyl 6-bromopyrazolo[1,5-a]pyridine-3-carboxylate
Formula: C₁₀H₉BrN₂O₂
MW: 269.095 g/mol
CAS Number: 55899-30-4

Order Now

Specification Test Results

Purety 97%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR