3-Bromo-1-methyl-1H-pyrazole ≥99.9%

N2011036-2 N2011036-3 N2011036-1
103535-468EA 101.54 USD
103535-468 103535-470 103536-370
3-Bromo-1-methyl-1H-pyrazole ≥99.9%
3-Bromo-1-methyl-1H-pyrazole
Formula: C4H5BrN2
MW: 161.0000 g/mol
MDL Number: MFCD11110668
CAS Number: 151049-87-5

Order Now

Specification Test Results

Purety 99.9%
Appearance Light-Yellow oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR