1,4-Dioxaspiro[4.5]decan-8-ol ≥97.0%

Synonyms: 4-Hydroxycyclohexan-1-one monoethylene ketal, 4-Hydroxycyclohexanone ethylene acetal

LJ7204-1 LJ7204-2 LJ7204-3
103536-352EA 23.08 USD
103536-352 103536-354 103536-356
1,4-Dioxaspiro[4.5]decan-8-ol ≥97.0%
1,4-Dioxaspiro[4.5]decan-8-ol
Formula: C₈H₁₄O₃
MW: 158.19 g/mol
MDL Number: MFCD00067003
CAS Number: 22428-87-1

Order Now

Specification Test Results

Purety 97.0%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR