N4-Benzoylcytosine ≥98.1%

Synonyms: N-(2-oxo-1,2-dihydropyrimidin-4-yl)benzamide

LJ7008-1 LJ7008-2 LJ7008-3
103536-346EA 12.31 USD
103536-346 103536-348 103536-350
N4-Benzoylcytosine ≥98.1%
N4-Benzoylcytosine
Formula: C₁₁H₉N₃O₂
MW: 215.21 g/mol
MDL Number: MFCD00239434
CAS Number: 26661-13-2

Order Now

Specification Test Results

Purety 98.1%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR