3-Oxabicyclo[3.1.0]hexane-2,4-dione ≥99.3%

Synonyms: Cyclopropane-1,2-dicarboxylic acid anhydride

LH1162-1 LH1162-2 LH1162-3
103535-466EA 30.77 USD
103535-466 103536-342 103536-344
3-Oxabicyclo[3.1.0]hexane-2,4-dione ≥99.3%
3-Oxabicyclo[3.1.0]hexane-2,4-dione
Formula: C5H4O3
MW: 112.08 g/mol
MDL Number: MFCD00126929
CAS Number: 5617-74-3

Order Now

Specification Test Results

Purety 99.3%
Appearance Light-Yellow crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR