{7-Methyl-7-azaspiro[3.5]nonan-2-yl}methanol ≥98.0%

LG1023-1 LG1023-2 LG1023-3
103535-460EA 115.38 USD
103535-460 103535-462 103535-464
{7-Methyl-7-azaspiro[3.5]nonan-2-yl}methanol ≥98.0%
{7-Methyl-7-azaspiro[3.5]nonan-2-yl}methanol
Formula: C₁₀H₁₉NO
CAS Number: 1363360-02-4

Order Now

Specification Test Results

Purety 98.0%
Appearance Colorless oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR