(4-Bromophenyl)acetic acid ≥97.0%

Synonyms: 4-Bromophenylacetic acid, 2-(4-bromophenyl)acetic acid

L2011003-1 L2011003-2 L2011003-3
103535-454EA 30.77 USD
103535-454 103535-456 103535-458
(4-Bromophenyl)acetic acid ≥97.0%
(4-Bromophenyl)acetic acid
Formula: (C₆H₄Br)CH₂COOH
MW: 215.05 g/mol
MDL Number: MFCD00004342
CAS Number: 1878-68-8

Order Now

Specification Test Results

Purety 97.0%
Appearance white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR