3-Bromo-α,α,α-trifluoro-5-nitrotoluene ≥99.5%

HT8102-1 HT8102-2 HT8102-3
103535-442EA 12.31 USD
103535-442 103535-444 103535-446
3-Bromo-α,α,α-trifluoro-5-nitrotoluene ≥99.5%
3-Bromo-α,α,α-trifluoro-5-nitrotoluene
Formula: C₇H₃BrF₃NO₂
MW: 270.01 g/mol
MDL Number: MFCD03788719
CAS Number: 630125-49-4

Order Now

Specification Test Results

Purety 99.5%
Appearance Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR