1-[2-(2-Aminoethoxy)ethyl]-2,5-dihydro-1H-pyrrole-2,5-dione trifluoroacetate ≥98.0%

Synonyms: 1-(2-(2-Aminoethoxy)ethyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate

HT8046-1 HT8046-2 HT8046-3
103535-436EA 230.77 USD
103535-436 103535-438 103535-440
1-[2-(2-Aminoethoxy)ethyl]-2,5-dihydro-1H-pyrrole-2,5-dione trifluoroacetate ≥98.0%
1-[2-(2-Aminoethoxy)ethyl]-2,5-dihydro-1H-pyrrole-2,5-dione trifluoroacetate
Formula: C₁₀H₁₃F₃N₂O₅
MW: 298.216 g/mol
MDL Number: MFCD27987223
CAS Number: 131274-17-4

Order Now

Specification Test Results

Purety 98.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR