2-(Trimethylsilyl)ethoxymethyl chloride ≥97.0%

H04161R-1 H04161R-2 H04161R-3
103535-430EA 23.08 USD
103535-430 103535-432 103535-434
2-(Trimethylsilyl)ethoxymethyl chloride ≥97.0%
2-(Trimethylsilyl)ethoxymethyl chloride
Formula: C₆H₁₅ClOSi
MDL Number: MFCD00009919
CAS Number: 76513-69-4
UN: 2920
ADR: 3,II

Order Now

Specification Test Results

Purety 97.0%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR