N-Carbobenzyloxy-L-alanine ≥97.0%

Synonyms: N-Cbz-L-alanine, Z-Ala-OH, (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid, N-Benzyloxycarbonyl-L-alanine, ((Benzyloxy)carbonyl)-L-alanine

93228-1 93228-2 93228-3
103535-424EA 46.15 USD
103535-424 103535-426 103535-428
N-Carbobenzyloxy-L-alanine ≥97.0%
N-Carbobenzyloxy-L-alanine
Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
MDL Number: MFCD00002640
CAS Number: 1142-20-7

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR