2-Chloro-4-iodo-6-(trifluoromethyl)pyridine ≥97.7%

90832-1 90832-2 90832-3
103536-324EA 46.15 USD
103536-324 103536-326 103536-328
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine ≥97.7%
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine
Formula: C₆H₂ClF₃IN
MDL Number: MFCD10699394
CAS Number: 205444-22-0

Order Now

Specification Test Results

Purety 97.7%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR