(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride ≥98.2%

Synonyms: cis-1-Benzyl-N3,4-dimethylpiperidin-3-amine dihydrochloride, cis-1-Benzyl-4-methyl-3-(methylamino)piperidine dihydrochloride

90363-1-1 90363-1-2 90363-1-3
103536-306EA 184.62 USD
103536-306 103536-308 103536-310
(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride ≥98.2%
(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride
Formula: C₁₄H₂₄Cl₂N₂
MW: 291.26 g/mol
MDL Number: MFCD09864744
CAS Number: 1062580-52-2

Order Now

Specification Test Results

Purety 98.2%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR