4,4'-Di-tert-butylbiphenyl ≥98.1%

Synonyms: 1-tert-butyl-4-(4-tert-butylphenyl)benzene, 4,4'-Di-tert-butyl-1,1'-biphenyl

90342-1 90342-2 90342-3
103535-422EA 30.77 USD
103535-422 103536-302 103536-304
4,4'-Di-tert-butylbiphenyl ≥98.1%
4,4'-Di-tert-butylbiphenyl
Formula: C₂₀H₂₆
MW: 266.43 g/mol
MDL Number: MFCD00008834
CAS Number: 1625-91-8

Order Now

Specification Test Results

Purety 98.1%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR