tert-Butoxybis(dimethylamino)methane ≥90.0%

Synonyms: Bredereck's reagent, 1-tert-Butoxy-N,N,N',N'-tetramethylmethylenenediamine, [(tert-butoxy)(dimethylamino)methyl]dimethylamine, tert-Butoxy bis(dimethylamino)methane

90107-1 90107-2 90107-3
103535-416EA 36.92 USD
103535-416 103535-418 103535-420
tert-Butoxybis(dimethylamino)methane ≥90.0%
tert-Butoxybis(dimethylamino)methane
Formula: C₉H₂₂N₂O
MW: 174.288 g/mol
MDL Number: MFCD00042858
CAS Number: 5815-08-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Purety 90.0%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR