cis-4-Aminocyclohexanol hydrochloride ≥97.0%

Synonyms: cis-4-Aminocyclohexan-1-ol hydrochloride, cis-4-Hydroxycyclohexylamine hydrochloride

90038-01-1 90038-01-2 90038-01-3
103535-410EA 30.77 USD
103535-410 103535-412 103535-414
cis-4-Aminocyclohexanol hydrochloride ≥97.0%
cis-4-Aminocyclohexanol hydrochloride
Formula: C₆H₁₄ClNO
MW: 151.63 g/mol
MDL Number: MFCD07366531
CAS Number: 56239-26-0

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR