2,6-Difluoro-3-(propane-1-sulfonamido)benzoic acid ≥98.3%

Synonyms: 2,6-difluoro-3-(propane-1-sulphonamido)benzoic acid, 2,6-Difluoro-3-(propylsulphonamido)benzoic acid

2013001-1 2013001-2 2013001-3
103535-398EA 30.77 USD
103535-398 103535-400 103535-402
2,6-Difluoro-3-(propane-1-sulfonamido)benzoic acid ≥98.3%
2,6-Difluoro-3-(propane-1-sulfonamido)benzoic acid
Formula: C₁₀H₁₁F₂NO₄S
MW: 279.26 g/mol
MDL Number: MFCD18157658
CAS Number: 1103234-56-5

Order Now

Specification Test Results

Purety 98.3%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR