2-Fluoropyridine ≥99.9%

130001-1 130001-2 130001-3
103535-392EA 32.31 USD
103535-392 103535-394 103535-396
2-Fluoropyridine ≥99.9%
2-Fluoropyridine
Formula: C₅H₄FN
MW: 97.09 g/mol
MDL Number: MFCD00006224
CAS Number: 372-48-5

Order Now

Specification Test Results

Purety 99.9%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR