Methyl-3-methyl-4-nitrobenzoate ≥97.0%

Synonyms: Methyl 3-methyl-4-nitrobenzoate

125929-1 125929-2 125929-3
103535-386EA 12.31 USD
103535-386 103535-388 103535-390
Methyl-3-methyl-4-nitrobenzoate ≥97.0%
Methyl 3-methyl-4-nitrobenzoate
Formula: C₉H₉NO₄
MW: 195.18 g/mol
MDL Number: MFCD00085640
CAS Number: 24078-21-5

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR