4,6-Dichloronicotinic acid ≥98.8%

Synonyms: 4,6-dichloropyridine-3-carboxylic acid

125920-1 125920-2 125920-3
103536-296EA 30.77 USD
103536-296 103536-298 103536-300
4,6-Dichloronicotinic acid ≥98.8%
4,6-Dichloronicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192.00 g/mol
MDL Number: MFCD00234146
CAS Number: 73027-79-9

Order Now

Specification Test Results

Purety 98.8%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR