4-Bromopyridinium chloride ≥99.1%

Synonyms: 4-Bromopyridine HCl, 4-Bromopyridine hydrochloride

124937-1 124937-2 124937-3
103535-376EA 30.77 USD
103535-376 103535-378 103535-380
4-Bromopyridinium chloride ≥99.1%
4-Bromopyridinium chloride
Formula: C₅H₅BrClN
MW: 194.46 g/mol
MDL Number: MFCD00012828
CAS Number: 19524-06-2

Order Now

Specification Test Results

Purety 99.1%
Appearance White crysta

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR