1,4-Dioxaspiro[4.5]decan-8-one ≥98.5%

124143-1 124143-2 124143-3
103535-370EA 30.77 USD
103535-370 103535-372 103535-374
1,4-Dioxaspiro[4.5]decan-8-one ≥98.5%
1,4-Dioxaspiro[4.5]decan-8-one
Formula: C₈H₁₂O₃
MW: 156.18 g/mol
MDL Number: MFCD00010214
CAS Number: 4746-97-8

Order Now

Specification Test Results

Purety 98.5%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR