Methyl-2-methylnicotinate ≥97.0%

Synonyms: methyl 2-methylpyridine-3-carboxylate, Methyl 2-methylnicotinate

123942-1 123942-2 123942-3
103535-364EA 30.77 USD
103535-364 103535-366 103535-368
Methyl-2-methylnicotinate ≥97.0%
Methyl-2-methylnicotinate
Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00051961
CAS Number: 65719-09-7

Order Now

Specification Test Results

Purety 97.0%
Appearance Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR