(Trifluoromethyl)trimethylsilane ≥97.0%

Synonyms: trimethyl(trifluoromethyl)silane

123806-1 123806-2 123806-3
103535-358EA 12.31 USD
103535-358 103535-360 103535-362
(Trifluoromethyl)trimethylsilane ≥97.0%
(Trifluoromethyl)trimethylsilane
Formula: C₄H₉F₃Si
MW: 142.2 g/mol
MDL Number: MFCD00145454
CAS Number: 81290-20-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Purety 97.0%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR