1-Boc-3-cyanopiperidine ≥97.0%

Synonyms: tert-butyl 3-cyanopiperidine-1-carboxylate, 1-Boc-3-cyanopiperidine, 3-Cyano-1-piperidinecarboxylic acid tert-Butyl ester, tert-Butyl 3-Cyano-1-piperidinecarboxylate, 1-Boc-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-cyanopiperidine

123513-1 123513-2 123513-3
103535-334EA 30.77 USD
103535-334 103535-336 103535-338
1-Boc-3-cyanopiperidine ≥97.0%
1-Boc-3-cyanopiperidine
Formula: C₁₁H₁₈N₂O₂
MW: 210.27 g/mol
MDL Number: MFCD01861222
CAS Number: 91419-53-3

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR