6-Hydroxy-2-naphthoic acid ≥99.3%

Synonyms: 6-hydroxynaphthalene-2-carboxylic acid

123372-1 123372-2 123372-3
103535-328EA 12.31 USD
103535-328 103535-330 103535-332
6-Hydroxy-2-naphthoic acid ≥99.3%
6-Hydroxy-2-naphthoic acid
Formula: C₁₁H₈O₃
MW: 188.18 g/mol
MDL Number: MFCD00060070
CAS Number: 16712-64-4

Order Now

Specification Test Results

Purety 99.3%
Appearance Light-Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR