Ethyl-2-chloropyrimidine-5-carboxylate ≥97.3%

Synonyms: Ethyl 2-chloropyrimidine-5-carboxylate

123364-1 123364-2 123364-3
103535-322EA 30.77 USD
103535-322 103535-324 103535-326
Ethyl-2-chloropyrimidine-5-carboxylate ≥97.3%
Ethyl-2-chloropyrimidine-5-carboxylate
Formula: C₇H₇ClN₂O₂
MW: 186.60 g/mol
MDL Number: MFCD09863164
CAS Number: 89793-12-4

Order Now

Specification Test Results

Purety 97.3%
Appearance Light-Yellow Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR